Anti-Cd8a Rat Monoclonal Antibody [clone: 53-6.7] (Cy5®)
Catalog # PRSI98-621
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD8 antigen, alpha chain
- Antigen symbol:Cd8a
- Clonality:Monoclonal
- Clone:[53-6.7]
- Conjugation:Cy5®
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Form:Liquid
- Gene ID:12525
- Antigen synonyms:T8|Ly-2|Lyt-2|Ly-35|Ly-B
- Modification:None
- Storage buffer:Supplied in PBS/NaN₃
- Storage temperature:Store vial at 2...8 °C
- Concentration:0,1 mg/ml
- Shipping temperature:20
- Immunogen:Spleen cells or thymocyte membranes
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: [53-6.7]
Conjugation: Cy5®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse