Order Entry
Italy
ContactUsLinkComponent
Anti-Cd8a Rat Monoclonal Antibody [clone: 53-6.7] (APC (Allophycocyanin))
Anti-Cd8a Rat Monoclonal Antibody [clone: 53-6.7] (APC (Allophycocyanin))
Catalog # PRSI98-618
Supplier:  ProSci Inc.
Anti-Cd8a Rat Monoclonal Antibody [clone: 53-6.7] (APC (Allophycocyanin))
Catalog # PRSI98-618
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8 antigen, alpha chain
  • Antigen symbol:
    Cd8a
  • Clonality:
    Monoclonal
  • Clone:
    [53-6.7]
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    12525
  • Antigen synonyms:
    T8|Ly-2|Lyt-2|Ly-35|Ly-B
  • Modification:
    None
  • Storage buffer:
    Supplied in PBS/NaN₃ and a stabilising agent
  • Storage temperature:
    Store vial at 2...8 °C
  • Concentration:
    0,1 mg/ml
  • Shipping temperature:
    20
  • Immunogen:
    Spleen cells or thymocyte membranes
  • Tested applications:
    Multiplex
  • Pack type:
    Vial
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: [53-6.7]
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse