Anti-GKN1 Rabbit Polyclonal Antibody
Catalog # PRSI14-797
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Gastrokine-1
- Antigen symbol:GKN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:56287
- Antigen synonyms:Protein CA11|AMP-18|GKN1|AMP18|CA11|18 kDa antrum mucosa protein|Gastrokine-1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 25-185 of human GKN1 (NP_062563.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GKN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat