Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # PRSI14-458
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:protein kinase, AMP-activated, beta 1 non-catalytic subunit
- Antigen symbol:PRKAB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5564
- Antigen synonyms:AMPK subunit beta-1|AMPK|HAMPKb|AMPK beta -1 chain|PRKAB1|AMPK beta 1|AMPKb
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human PRKAB1 (NP_006244.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PRKAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat