Anti-RAB34 Rabbit Polyclonal Antibody
PRSI13-585
New Product
- Antibody type:Primary
- Antigen name:RAB34, member RAS oncogene family
- Antigen symbol:RAB34
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:83871
- Antigen synonyms:ras-related protein Rah|RAB39|ras-related protein Rab-34|RAH|ras-related protein Rab-39|NARR|nine amino-acid residue-repeats
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-259 of human RAB34 (NP_114140.4)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: RAB34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat