Anti-SLC28A3 Rabbit Polyclonal Antibody
PRSI13-579
- Antibody type:Primary
- Antigen name:solute carrier family 28 member 3
- Antigen symbol:SLC28A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:64078
- Antigen synonyms:solute carrier family 28 (sodium-coupled nucleoside transporter)|concentrative Na(+)-nucleoside cotransporter 3|member 3|concentrative Na+-nucleoside cotransporter|CNT3|solute carrier family 28 (concentrative nucleoside transporter)|solute carrier family 28 member 3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 612-691 of human SLC28A3 (NP_071410.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SLC28A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse