Order Entry
Italy
ContactUsLinkComponent
 
Anti-PRKAG1 Rabbit Polyclonal Antibody
  PRSI62-619
New Product
undefined
Anti-PRKAG1 Rabbit Polyclonal Antibody
  PRSI62-619
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    5'-AMP-activated protein kinase subunit gamma-1
  • Antigen symbol:
    PRKAG1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    265-295 aa
  • Form:
    Liquid
  • Gene ID:
    5571
  • Modification:
    None
  • Molecular weight:
    38 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Shipping temperature:
    4 °C
  • Immunogen:
    This AMPK gamma (PRKAG1) antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 265-295 amino acids from the central region of human AMPK gamma (PRKAG1).
  • Purification:
    This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
  • Pk:
    400 µl

 

 

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: PRKAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 265-295
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse