Anti-MTMR4 Rabbit Polyclonal Antibody
PRSI23-165
- Antibody type:Primary
- Antigen name:Myotubularin-related protein 4
- Antigen symbol:MTMR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:9110
- Antigen synonyms:KIAA0647|FYVE domain-containing dual specificity protein phosphatase 2|MTMR4|ZFYVE11|Myotubularin-related protein 4|Zinc finger FYVE domain-containing protein 11|FYVE-DSP2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 570-820 of human MTMR4 (NP_004678.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: MTMR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat