Anti-PLA2G2D Rabbit Polyclonal Antibody
PRSI22-408
- Antibody type:Primary
- Antigen name:Group IID secretory phospholipase A2
- Antigen symbol:PLA2G2D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:26279
- Antigen synonyms:SPLASH|GIID sPLA2|Secretory-type PLA|stroma-associated homolog|Phosphatidylcholine 2-acylhydrolase 2D|PLA2G2D|PLA2IID|Group IID secretory phospholipase A2|sPLA2-IID
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 21-145 of human PLA2G2D (NP_036532.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: PLA2G2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat