Anti-INPP5E Rabbit Polyclonal Antibody
Catalog # PRSI18-080
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Inositol polyphosphate-5-phosphatase E
- Antigen symbol:INPP5E
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:56623
- Antigen synonyms:CPD4|MORMS|phosphatidylinositol polyphosphate 5-phosphatase type IV|pharbin|JBTS1|CORS1|PPI5PIV|72 kDa inositol polyphosphate 5-phosphatase|phosphatidylinositol-4,5-bisphosphate 5-phosphatase
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 510-630 of human INPP5E (NP_063945,2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: INPP5E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat