Anti-PI4KB Rabbit Polyclonal Antibody
PRSI16-595
New Product
- Antibody type:Primary
- Antigen name:Phosphatidylinositol 4-kinase beta
- Antigen symbol:PI4KB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:5298
- Antigen synonyms:Phosphatidylinositol 4-kinase beta|PIK4CB|PtdIns 4-kinase beta|PI4K92|PI4KB|PI4K-beta|PI4Kbeta|NPIK
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 560-801 of human PI4KB (NP_001185702)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: PI4KB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat