Anti-NR2F1 Rabbit Polyclonal Antibody
Catalog # PRSI57-566
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:COUP transcription factor 1
- Antigen symbol:NR2F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:48-76 aa
- Form:Liquid
- Gene ID:7025
- Modification:None
- Molecular weight:46 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:This NR2F1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 48-76 amino acids from the N-terminal region of human NR2F1
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: NR2F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 48-76
Host: Rabbit
Isotype: Ig
Reactivity: Human