Anti-ZNF429 Rabbit Polyclonal Antibody
Catalog # PRSI56-742
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Zinc finger protein 429
- Antigen symbol:ZNF429
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino acid range = 646-674
- Form:Liquid
- Gene ID:Q86V71
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:78 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ZNF429 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 646-674 amino acids from the C-terminal region of human ZNF429
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
ZNF429 may be involved in transcriptional regulation.
- For WB starting dilution is: 1:1000
Type: Primary
Antigen: ZNF429
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human