Anti-CCNL2 Rabbit Polyclonal Antibody
PRSI56-397
New Product
- Antibody type:Primary
- Antigen name:Cyclin-L2
- Antigen symbol:CCNL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino acid range = 338-366
- Form:Liquid
- Gene ID:Q96S94
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:58 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CCNL2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 338-366 amino acids from the Central region of human CCNL2
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
CCNL2 is a transcriptional regulator which participates in regulating the pre-mRNA splicing process. Also modulates the expression of critical apoptotic factor, leading to cell apoptosis.
- For WB starting dilution is: 1:1000
Type: Primary
Antigen: CCNL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human