Order Entry
Italy
ContactUsLinkComponent
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Catalog # PRSI76-362
Supplier:  ProSci Inc.
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Catalog # PRSI76-362
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD4 molecule
  • Antigen symbol:
    CD4
  • Clonality:
    Monoclonal
  • Clone:
    RPA-T4
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P01730
  • Antigen synonyms:
    CD4
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    25 Tests

Specifications

About this item

The RPA-T4 monoclonal antibody specifically binds to the CD4 receptor for the human immunodeficiency virus (HIV). CD4 is a 59 kDa single-chain transmembrane glycoprotein that expressed on the surface of most of the thymocytes, T-helper cells, and in low levels on monocytes and macrophages. CD4 is a co-receptor in the antigen-induced T cell activation, together with the MHC class II. The RPA-T4 antibody is capable of blocking HIV binding and inhibiting syncytium formation, by binding to the D1 domain of the CD4 antigen. The OKT4 and the RPA-T4 monoclonal antibodies recognize different epitopes of CD4 and they do not exhibit cross-block binding.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human