Anti-H2-Ab1 Rat Monoclonal Antibody [clone: M5/114.15.2]
Catalog # PRSI77-190
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:histocompatibility 2, class II antigen A, beta 1
- Antigen symbol:H2-Ab1
- Clonality:Monoclonal
- Clone:M5/114.15.2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:P14483
- Antigen synonyms:H2-Ab1
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Tested applications:Functional, Electron Microscopy
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: H2-Ab1
Clonality: Monoclonal
Clone: M5/114.15.2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse