Order Entry
Italy
ContactUsLinkComponent
Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9] (APC (Allophycocyanin))
Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9] (APC (Allophycocyanin))
Catalog # PRSI76-151
Supplier:  ProSci Inc.
Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9] (APC (Allophycocyanin))
Catalog # PRSI76-151
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    integrin, alpha X (complement component 3 receptor 4 subunit)
  • Antigen symbol:
    ITGAX
  • Clonality:
    Monoclonal
  • Clone:
    3.9
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P20702
  • Antigen synonyms:
    ITGAX
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    25 Tests

Specifications

About this item

The 3.9 monoclonal antibody specifically binds to the human adhesion glycoprotein CD11c, a 150 kDa integrin alpha chain also known as integrin alpha X. It is expressed on macrophages, granulocytes, monocytes, dendritic cells, natural killer cells, and subsets of B and T lymphocytes. The CD11c/CD18 complex associates with the iC3b, fibrinogen and ICAM-1 and has an important function in leukocyte adhesion.

Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: 3.9
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human