Anti-CD1A Mouse Monoclonal Antibody [clone: HI149] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI76-084
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD1a molecule
- Antigen symbol:CD1A
- Clonality:Monoclonal
- Clone:HI149
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P06126
- Antigen synonyms:CD1A
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:25 Tests
Specifications
About this item
The HI149 monoclonal antibody specifically reacts with human CD1a, a 49 kDa type I membrane glycoprotein. It is expressed on dendritic cells, Langerhans cells, and cortical thymocytes. CD1a plays a role in antigen presentation and ,like MHC Class I, it non-covalently associates with Beta2-microglobulin.
Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: HI149
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human