Order Entry
Italy
ContactUsLinkComponent
Anti-Cd8a Rat Monoclonal Antibody [clone: 53-6.7] (APC (Allophycocyanin))
Anti-Cd8a Rat Monoclonal Antibody [clone: 53-6.7] (APC (Allophycocyanin))
Catalog # PRSI76-697
Supplier:  ProSci Inc.
Anti-Cd8a Rat Monoclonal Antibody [clone: 53-6.7] (APC (Allophycocyanin))
Catalog # PRSI76-697
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8 antigen, alpha chain
  • Antigen symbol:
    Cd8a
  • Clonality:
    Monoclonal
  • Clone:
    53-6.7
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P01731
  • Antigen synonyms:
    Cd8a
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,025 mg

Specifications

About this item

Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse