Anti-EIF1B Rabbit Polyclonal Antibody
PRSI58-069
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 1b
- Antigen symbol:EIF1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:84 - 113 aa
- Form:Liquid
- Gene ID:10289
- Modification:None
- Molecular weight:13 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This EIF1B antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 84-113 amino acids from the C-terminal region of human EIF1B
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: EIF1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 84-113
Host: Rabbit
Isotype: Ig
Reactivity: Human