Anti-SLC1A2 Rabbit Polyclonal Antibody
Catalog # PRSI13-320
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 1 member 2
- Antigen symbol:SLC1A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6506
- Antigen synonyms:EAAT2|solute carrier family 1 (glial high affinity glutamate transporter)|HBGT|sodium-dependent glutamate/aspartate transporter 2|GLT-1|member 2|EIEE41|glutamate/aspartate transporter II|excitatory amino acid transporter 2|excitotoxic amino acid transporter 2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 495-574 of human SLC1A2 (NP_004162.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC1A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat