Anti-R1450 Recombinant Antibody
PRSI10-243
New Product
- Antibody type:Primary
- Clonality:Recombinant
- Conjugation:Unconjugated
- Antigen synonyms:anti-ERBB2 (250A/TP-A-02)/Rat sp.G2b lambda|anti-CD3E (26/II/6-1.2)|TPBs03|Musmus G2a kappa
- Modification:None
- Storage buffer:PBS buffer pH 7,5
- Storage temperature:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Mus musculus,Rattus sp,Hybrid/CD3E/ERBB2/CD340 [Homo sapiens]
- Purification:>95%
- Pack type:Vial
- Pk:0,1 mg
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a-kappa/G2b-lambda
Reactivity: