Anti-RPS27A Rabbit Polyclonal Antibody
Catalog # PRSI18-452
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal protein S27a
- Antigen symbol:RPS27A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:6233
- Antigen synonyms:UBCEP1|UBCEP80|S27A|UBA80|CEP80|Ubiquitin carboxyl extension protein 80UBC|Ubiquitin-40S ribosomal protein S27a|HEL112
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-156 of human RPS27A (NP_001170884.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RPS27A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat