Anti-ACE Rabbit Polyclonal Antibody
Catalog # PRSI63-208
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Angiotensin-converting enzyme
- Antigen symbol:ACE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:1274-1306 aa
- Form:Liquid
- Gene ID:1636
- Modification:None
- Molecular weight:150 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ACE antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1274-1306 amino acids from the C-terminal region of human ACE.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ACE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 1274-1306
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse