Anti-SRSF2 Rabbit Polyclonal Antibody
PRSI61-513
- Antibody type:Primary
- Antigen name:Serine/arginine-rich splicing factor 2
- Antigen symbol:SRSF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:76 - 105 aa
- Form:Liquid
- Gene ID:6427
- Modification:None
- Molecular weight:25 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SFRS2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 76 - 105 amino acids from the Central region of human SFRS2.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: SRSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 76-105
Host: Rabbit
Isotype: Ig
Reactivity: Human