Order Entry
Italy
ContactUsLinkComponent
Anti-USP7 Rabbit Polyclonal Antibody
Anti-USP7 Rabbit Polyclonal Antibody
Catalog # PRSI61-101
Supplier:  ProSci Inc.
undefined
Anti-USP7 Rabbit Polyclonal Antibody
Catalog # PRSI61-101
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin carboxyl-terminal hydrolase 7
  • Antigen symbol:
    USP7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Epitope:
    1060 - 1090 aa
  • Form:
    Liquid
  • Gene ID:
    7874
  • Modification:
    None
  • Molecular weight:
    128 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This USP7 (HAUSP) antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 1060 - 1090 amino acids from the C-terminal region of human USP7 (HAUSP).
  • Purification:
    This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
  • Pk:
    400 µl

Specifications

About this item

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: USP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 1060-1090
Host: Rabbit
Isotype: Ig
Reactivity: Human