Anti-GTF3C3 Rabbit Polyclonal Antibody
Catalog # PRSI60-499
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:General transcription factor 3C polypeptide 3
- Antigen symbol:GTF3C3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:47 - 76 aa
- Form:Liquid
- Gene ID:9330
- Modification:None
- Molecular weight:101 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This GTF3C3 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 47 - 76 amino acids from the N-terminal region of human GTF3C3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: GTF3C3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 47-76
Host: Rabbit
Isotype: Ig
Reactivity: Human