Order Entry
Italy
ContactUsLinkComponent
Anti-C5AR1 Recombinant Antibody [Clone: 32-G1]
Anti-C5AR1 Recombinant Antibody [Clone: 32-G1]
Catalog # PRSI24-780
Supplier:  ProSci Inc.
Anti-C5AR1 Recombinant Antibody [Clone: 32-G1]
Catalog # PRSI24-780
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    complement C5a receptor 1
  • Antigen symbol:
    C5AR1
  • Clonality:
    Recombinant
  • Clone:
    32-G1
  • Conjugation:
    Unconjugated
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    728
  • Antigen synonyms:
    C5a anaphylatoxin chemotactic receptor|C5a-R|32G-1|C5a anaphylatoxin chemotactic receptor 1|C5aR|CD88|pC5aR
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was raised by immunizing mice a synthetic C5aR C-terminal phosphopeptide that contained phosphoserine instead of serine at position 334 and also at the neighboring phosphorylation sites Ser-332 and Ser-338
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: complement C5a receptor 1
Clonality: Recombinant
Clone: 32-G1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human