Anti-Slc3a2 Recombinant Antibody [Clone: RL388]
Catalog # PRSI24-682
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 3 (activators of dibasic and neutral amino acid transport), member 2
- Antigen symbol:Slc3a2
- Clonality:Recombinant
- Clone:RL388
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Gene ID:17254
- Antigen synonyms:RL-388|4F2|4F2hc|Solute carrier family 3 member 2|4F2 cell-surface antigen heavy chain|SLC3A2
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising rats with an IL 2-producing subline of the mouse T lymphoma EL4
- Tested applications:Stimulation
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: solute carrier family 3 (activators of dibasic and neutral amino acid transport), member 2
Clonality: Recombinant
Clone: RL388
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse