Order Entry
Italy
ContactUsLinkComponent
Anti-TAG-72 Recombinant Antibody [Clone: Minretumomab (CC49 )]
Anti-TAG-72 Recombinant Antibody [Clone: Minretumomab (CC49 )]
Catalog # PRSI24-642
Supplier:  ProSci Inc.
Anti-TAG-72 Recombinant Antibody [Clone: Minretumomab (CC49 )]
Catalog # PRSI24-642
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Clone:
    Minretumomab (CC49 )
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    Tumor-associated antigen 72|TAG72|CA 72-4|Tumor-associated glycoprotein 72
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was raised by immunising BALB/c mice with TAG-72 purified from LS-174T human carcinoma xenograft
  • Tested applications:
    RIA, therapeutics
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: TAG-72
Clonality: Recombinant
Clone: Minretumomab (CC49 )
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human