Anti-GM2 Recombinant Antibody [Clone: L55-81]
Catalog # PRSI24-512
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:L55-81
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Isotype:IgM kappa
- Cross adsorption:No
- Form:Liquid
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanoma
- Tested applications:TLC Depletion
- Pack type:Vial
- Pk:0,05 mg
Specifications
About this item
Type: Primary
Antigen: GM2
Clonality: Recombinant
Clone: L55-81
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgM kappa
Reactivity: Human