Anti-RRM2 Recombinant Antibody [Clone: SAIC-30C-18]
Catalog # PRSI24-328
Nome del fornitore: ProSci Inc.
Specifiche
- Antibody type:Primary
- Antigen name:ribonucleotide reductase regulatory subunit M2
- Antigen symbol:RRM2
- Clonality:Recombinant
- Clone:SAIC-30C-18
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:6241
- Antigen synonyms:RR2|ribonucleotide reductase regulatory subunit M2|R2|Ribonucleoside-diphosphate reductase subunit M2|RR2M
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:The KLH-conjugated peptide "IEQEFLTEALPVK" derived from Ribonucleoside-diphosphate reductase subunit M2 (RRM2). RRM2 provides precursors required for DNA synthesis
- Tested applications:ImmunoMRM
- Pack type:Vial
- Pk:0,2 mg
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: ribonucleotide reductase regulatory subunit M2
Clonality: Recombinant
Clone: SAIC-30C-18
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human