Order Entry
Italy
ContactUsLinkComponent
Anti-null Recombinant Antibody [Clone: SAIC-13B-19]
Anti-null Recombinant Antibody [Clone: SAIC-13B-19]
Catalog # PRSI24-310
Supplier:  ProSci Inc.
Anti-null Recombinant Antibody [Clone: SAIC-13B-19]
Catalog # PRSI24-310
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    null
  • Clonality:
    Recombinant
  • Clone:
    SAIC-13B-19
  • Conjugation:
    Unconjugated
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    2190
  • Antigen synonyms:
    ANXA1 protein|hCG_17305
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Peptide "AAYLQETGKPLDETLK" derived from Annexin A1 conjugated to KLH
  • Tested applications:
    ImmunoMRM
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: null
Clonality: Recombinant
Clone: SAIC-13B-19
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human