Order Entry
Italy
ContactUsLinkComponent
Anti-TAG-72 Recombinant Antibody [Clone: B72.3 (Satumomab)]
Anti-TAG-72 Recombinant Antibody [Clone: B72.3 (Satumomab)]
Catalog # PRSI24-276
Supplier:  ProSci Inc.
Anti-TAG-72 Recombinant Antibody [Clone: B72.3 (Satumomab)]
Catalog # PRSI24-276
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Clone:
    B72.3 (Satumomab)
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    TAG-72
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Tumour-associated glycoprotein 72 is a glycoprotein found on the surface of many cancer cells, including ovary, breast, colon, and pancreatic cells
  • Tested applications:
    Imaging
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: TAG-72
Clonality: Recombinant
Clone: B72.3 (Satumomab)
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human