Anti-CD4 Recombinant Antibody [Clone: YNB46.1.8 (Campath-9H)]
Catalog # PRSI24-198
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:HGNC:HGNC:1678
- Antigen symbol:CD4
- Clonality:Recombinant
- Clone:YNB46.1.8 (Campath-9H)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Cross adsorption:No
- Form:Liquid
- Gene ID:920
- Antigen synonyms:Cluster of differentiation 4|T-cell surface antigen T4/Leu-3|T-cell surface glycoprotein CD4|Leu-3/T4
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Rat T-cell line (NB2) transfected with the human CD4
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: HGNC:HGNC:1678
Clonality: Recombinant
Clone: YNB46.1.8 (Campath-9H)
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human