Anti-PRMT5 Recombinant Antibody [Clone: RAB-C136]
Catalog # PRSI24-839
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:protein arginine methyltransferase 5
- Antigen symbol:PRMT5
- Clonality:Recombinant
- Clone:RAB-C136
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:Fab fragment kappa
- Cross adsorption:No
- Form:Liquid
- Gene ID:10419
- Antigen synonyms:anti-PRMT5-RAB-C136|Protein arginine N-methyltransferase 5|Histone-arginine N-methyltransferase PRMT5|SKB1 homolog|SKB1Hs|48F3_10|PRMT5|Jak-binding protein 1|72 kDa ICln-binding protein|Shk1 kinase-binding protein 1 homolog
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT5 protein under non-denaturing conditions
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: protein arginine methyltransferase 5
Clonality: Recombinant
Clone: RAB-C136
Conjugation: Unconjugated
Host: Recombinant
Isotype: Fab fragment kappa
Reactivity: Human