Order Entry
Italy
ContactUsLinkComponent
Anti-CD53 Recombinant Antibody [Clone: 161-2 (53/2)]
Anti-CD53 Recombinant Antibody [Clone: 161-2 (53/2)]
Catalog # PRSI24-757
Supplier:  ProSci Inc.
Anti-CD53 Recombinant Antibody [Clone: 161-2 (53/2)]
Catalog # PRSI24-757
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD53 molecule
  • Antigen symbol:
    CD53
  • Clonality:
    Recombinant
  • Clone:
    161-2 (53/2)
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    963
  • Antigen synonyms:
    Leukocyte surface antigen CD53|Tetraspanin-25|Tspan-25|Cell surface glycoprotein CD53
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was raised by immunizng mice with stimulated human leukocytes
  • Tested applications:
    Functional Assays
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: CD53 molecule
Clonality: Recombinant
Clone: 161-2 (53/2)
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2a kappa
Reactivity: Human