Anti-TAG-72 Recombinant Antibody [Clone: Minretumomab (CC49 )]
Catalog # PRSI24-641
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:Minretumomab (CC49 )
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Tumor-associated antigen 72|TAG72|CA 72-4|Tumor-associated glycoprotein 72
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising BALB/c mice with TAG-72 purified from LS-174T human carcinoma xenograft
- Tested applications:RIA,Therapeutics
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary,Biosimilars
Antigen: TAG-72
Clonality: Recombinant
Clone: Minretumomab (CC49 )
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human