Anti-IL16 Recombinant Antibody [Clone: 14.1]
Catalog # PRSI24-577
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:interleukin 16
- Antigen symbol:IL16
- Clonality:Recombinant
- Clone:14.1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:3603
- Antigen synonyms:interleukin 16|IL16|NPRprIL-16|LCF
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:The antibody was raised against IL-16 using a synthetic peptide corresponding to the predicted amino acid sequence of a hydrophilic region of IL-16, conjugated to keyhole limpet hemocyanin
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: interleukin 16
Clonality: Recombinant
Clone: 14.1
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2a kappa
Reactivity: Human