Order Entry
Italy
ContactUsLinkComponent
 
Anti-GM2 Recombinant Antibody [Clone: L55-81]
  PRSI24-513
Anti-GM2 Recombinant Antibody [Clone: L55-81]
  PRSI24-513

 

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Clone:
    L55-81
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanom
  • Tested applications:
    TLCDepletion
  • Pack type:
    Vial
  • Pk:
    0,2 mg

 

 

Type: Primary
Antigen: GM2
Clonality: Recombinant
Clone: L55-81
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human