Order Entry
Italy
ContactUsLinkComponent
 
Anti-SELL Recombinant Antibody [Clone: DREG-56]
  PRSI24-439
Anti-SELL Recombinant Antibody [Clone: DREG-56]
  PRSI24-439

 

  • Antibody type:
    Primary
  • Antigen name:
    selectin L
  • Antigen symbol:
    SELL
  • Clonality:
    Recombinant
  • Clone:
    DREG-56
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Chimpanzee,
    Human,
    Bovine
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    6402
  • Antigen synonyms:
    LAM-1|LECAM1|Lymph node homing receptor|Lymphocyte surface MEL-14 antigen|MEL-14|Leukocyte adhesion molecule 1|Ly-22|LECAM-1|CD62 antigen-like family member L|Lymphocyte antigen 22|L-selectin|Leukocyte-endothelial cell adhesion molecule 1
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was produced by immunizing mice with the concentrated supernatant generated after centrifugation of human peripheral blood leukocyte
  • Tested applications:
    Blocking
  • Pack type:
    Vial
  • Pk:
    0,2 mg

 

 

Type: Primary
Antigen: selectin L
Clonality: Recombinant
Clone: DREG-56
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human;Chimpanzee;Bovine