Order Entry
Italy
ContactUsLinkComponent
 
Anti-Pig IgG (H+L) Goat Polyclonal Antibody (CF®583R)
 :  Biotium
New Product
 
Anti-Pig IgG (H+L) Goat Polyclonal Antibody (CF®583R)
 :  Biotium
New Product

 

  • Catalog No:
  • 20907-50UL
  • 20907-500UL
  • Antigen name:
  • Pig IgG
  • Pig IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 583R
  • CF® 583R
  • Reactivity:
  • Pig
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Concentration:
  • 2 mg/ml
  • 2 mg/ml
  • Cross adsorption:
  • Not cross-adsorbed
  • Not cross-adsorbed
  • Storage temperature:
  • −20 °C
  • −20 °C

 

Frequently Bought Together