Riboflavine (vitamin B2)
Glass bottle for solids, 0,12 mg
Catalog # EDQMY0000757
Supplier: EDQM
Jump to:
Product Details & Documents
Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G
- CAS Number:83-88-5
- Formula:C₁₇H₂₀N₄O₆
- MDL Number:MFCD00005022
- Molecular Weight:376.37 g/mol
- Merck Index:12,08367
- Melting Pt:280 °C
- Storage Temperature:Ambient
- EINECS:201-507-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:0,12 mg



