
3-Thiophenecarbonitrile 94%
ACRO263160010
:
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5595 to 1.5615 (20°C, 589 nm)
- GC:≥92 %
:
:109.15 g/mol
:82 °C (10 mmHg)
:1,1956 g/cm³ (20 °C)
:216-687-7
:6.1,III
:GHS07
:1641-09-4
:C₅H₃NS
:MFCD00151852