Order Entry
Italy
ContactUsLinkComponent
Methyl red hydrochloride in ACS Reagent grade water, Sigma-Aldrich®
Methyl red hydrochloride in ACS Reagent grade water, Sigma-Aldrich®
Catalog # 861235-25G
CAS Number:  
undefined
Methyl red hydrochloride in ACS Reagent grade water, Sigma-Aldrich®
Catalog # 861235-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance (Colour):
    Light Blue to Very Dark Blue and Light Purple to Very Dark Purple
  • Appearance (Form):
    Powder or Crystals
  • Infrared spectrum:
    Conforms to Structure
  • Clarity of Solution
    c = 0.1%; Ethanol:
    Conforms
  • Visual Interval
    pH 4.2:
    Pink
  • Visual Interval
    pH 5.5:
    Orange
  • Visual Interval
    pH 6.2:
    Yellow
  • Meets ACS Requirements:
    Current ACS Specification

Specification Test Results

About this item

 :
 :305.76 g/mol
Melting Pt:175 °C
 :M-Clarity™ quality level = MQ200
EINECS:264-190-9
CAS Number:63451-28-5
Formula:C₁₅H₁₆ClN₃O₂
MDL Number:MFCD00012614