Ethylene di(acetate) 99%
:111-55-7
:
:C₆H₁₀O₄
:MFCD00008718
:146.14 g/mol
:13,03833
:186…187 °C (760 mmHg)
:–41 °C
:1.12 g/cm³
:82 °C
:Ambient
:203-881-1
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4142 to 1.4162 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥98.5 %
- Color scale:≤10 APHA
- Acidity (CH3COOH):≤0.1 %
Frequently Bought Together