Order Entry
Italy
ContactUsLinkComponent
Anti-CD3E Mouse Monoclonal Antibody [clone: SK7] (CF®660C)
Anti-CD3E Mouse Monoclonal Antibody [clone: SK7] (CF®660C)
Supplier:  Biotium
Certificates
Anti-CD3E Mouse Monoclonal Antibody [clone: SK7] (CF®660C)
Supplier:  Biotium

Specifications

  • Catalog No:
  • P002-660C-125
  • P002-660C-500
  • Antigen symbol:
  • CD3E
  • CD3E
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 660C
  • CF® 660C
  • Clone:
  • SK7
  • SK7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 916
  • 916
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Human thymocytes
  • Human thymocytes
  • Molecular weight:
  • 23 kDa
  • 23 kDa
  • Concentration:
  • 200 µg/ml
  • 200 µg/ml
  • Storage buffer:
  • PBS, 0,1% BSA, 0,05% azide
  • PBS, 0,1% BSA, 0,05% azide
  • Storage temperature:
  • Store at 2...8 °C, protect fluorescent conjugates from light
  • Store at 2...8 °C, protect fluorescent conjugates from light
  • Shipping temperature:
  • RT
  • RT
  • Tested applications:
  • Flow (surface) (verified)
  • Flow (surface) (verified)

Specifications

Frequently Bought Together