Order Entry
Italy
ContactUsLinkComponent
 
Anti- Llama Polyclonal Antibody (CF®583R)
 :  Biotium
New Product
 
Anti- Llama Polyclonal Antibody (CF®583R)
 :  Biotium
New Product

 

  • Catalog No:
  • 20909-500UL
  • 20909-50UL
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Rabbit IgG
  • Rabbit IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Host:
  • Llama
  • Llama
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Concentration:
  • 2 mg/ml
  • 2 mg/ml
  • Cross adsorption:
  • Not cross-adsorbed
  • Not cross-adsorbed
  • Storage temperature:
  • Store at −20 °C, protected from light.
  • Store at −20 °C, protected from light.

 

Frequently Bought Together