About this item
CD4 (clone SK3) is a mouse monoclonal antibody that recognizes the extracellular domain of CD4, a membrane glycoprotein of T lymphocytes. This antibody belongs to the Biotium choice list of select antibodies that have been validated and optimized in-house for optimal performance. The antibody is available conjugated to CF® Dyes and Astral™ tandem dyes. They are supplied in PBS, 0,1% BSA, 0,05% azide.
- Recommended concentration for flow cytometry: 5 µl per million cells/0,1 ml
- Protect fluorescent conjugates from light
CD4 interacts with major histocompatibility complex class II antigens and is also a receptor for the human immunodeficiency virus. This protein is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of the central nervous system. The majority of peripheral T-cell lymphomas are derived from the T-helper/regulatory cell subset so that most mature T-cell neoplasms are CD4/CD8.
Anti-CD4 is used in the immunohistochemical staining of lymphoproliferative disorders to evaluate tumors with CD4 aberrant expression.
Type: Primary
Antigen: CD4
Clonality: monoclonal
Clone: SK3
Conjugation: CF®660C
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Specifications
- Catalog No:
- P001-660C-125
- P001-660C-500
- Antigen symbol:
- CD4
- CD4
- Antigen name:
- CD4
- CD4
- Antigen synonyms:
- L3T4|T cell differentiation antigen L3T4|Leu3|T-cell surface antigen T4/Leu-3|p55|T cell antigen T4/LEU3|Lymphocyte antigen CD4|Ly-4|T-cell surface glycoprotein CD4
- L3T4|T cell differentiation antigen L3T4|Leu3|T-cell surface antigen T4/Leu-3|p55|T cell antigen T4/LEU3|Lymphocyte antigen CD4|Ly-4|T-cell surface glycoprotein CD4
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 660C
- CF® 660C
- Clone:
- SK3
- SK3
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 920
- 920
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human peripheral blood T cells
- Human peripheral blood T cells
- Molecular weight:
- 55 kDa
- 55 kDa
- Concentration:
- 5 µl per million cells/0,1 ml
- 5 µl per million cells/0,1 ml
- Storage temperature:
- Store at 2...8 °C, protect fluorescent conjugates from light
- Store at 2...8 °C, protect fluorescent conjugates from light
- Shipping temperature:
- RT
- RT