Phorbol-12-myristate-13-acetate ≥99% (by HPLC, TLC)
Catalog # ENZOBMLPE1600005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:616.836 g/mol
Storage Temperature:Freezer
CAS Number:16561-29-8
Formula:C₃₆H₅₆O₈
MDL Number:MFCD00036736